ContactUsLinkComponent
Mouse IL-22 RA2/IL-22BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of mouse IL-22 RA2/IL-22BP in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6190-96
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-22 RA2/IL-22BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of mouse IL-22 RA2/IL-22BP in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6190-96
Supplier:  ANTIBODIES.COM