ContactUsLinkComponent
Human Cytochrome P450 2B6/CYP2B6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Cytochrome P450 2B6/CYP2B6 in tissue homogenates, cell lysates or other biological flui 1 * 96 Tests
Catalog # ANTIA82311-96
Supplier:  ANTIBODIES.COM
New Product
Human Cytochrome P450 2B6/CYP2B6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Cytochrome P450 2B6/CYP2B6 in tissue homogenates, cell lysates or other biological flui 1 * 96 Tests
Catalog # ANTIA82311-96
Supplier:  ANTIBODIES.COM