ContactUsLinkComponent
Rat VDAC1/Porin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat VDAC1/Porin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA82394-96
Supplier:  ANTIBODIES.COM
New Product
Rat VDAC1/Porin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat VDAC1/Porin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA82394-96
Supplier:  ANTIBODIES.COM