ContactUsLinkComponent
Rat Mannan Binding Lectin/MBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Mannan Binding Lectin/MBL in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 96 Tests
  ANTIA7898-96
New Product
Rat Mannan Binding Lectin/MBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Mannan Binding Lectin/MBL in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 96 Tests
  ANTIA7898-96