ContactUsLinkComponent
Rat Estrogen Related Receptor gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of rat Estrogen Related Receptor gamma in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA79303-96
Supplier:  ANTIBODIES.COM
New Product
Rat Estrogen Related Receptor gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of rat Estrogen Related Receptor gamma in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA79303-96
Supplier:  ANTIBODIES.COM