ContactUsLinkComponent
Human HIV (HIV1 + HIV2 antibody + p24 antigen) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the i n vitro qualitative determination of human HIV (HIV1 + HIV2 antibody + p24 antigen) in serum, plasma, tissue homogena 1 * 96 Tests
Catalog # ANTIA73730-96
Supplier:  ANTIBODIES.COM
New Product
Human HIV (HIV1 + HIV2 antibody + p24 antigen) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the i n vitro qualitative determination of human HIV (HIV1 + HIV2 antibody + p24 antigen) in serum, plasma, tissue homogena 1 * 96 Tests
Catalog # ANTIA73730-96
Supplier:  ANTIBODIES.COM