ContactUsLinkComponent
Human Integrin Linked Kinase 1/ILK-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Integrin Linked Kinase 1/ILK-1 in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA77597-96
Supplier:  ANTIBODIES.COM
New Product
Human Integrin Linked Kinase 1/ILK-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qu antitative determination of human Integrin Linked Kinase 1/ILK-1 in serum, plasma, tissue homogenates, and other biol 1 * 96 Tests
Catalog # ANTIA77597-96
Supplier:  ANTIBODIES.COM