Human alpha II Spectrin Breakdown Product 120 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human alpha II Spectrin Breakdown Product 120 in serum, plasma, tissue homogenat 1 * 96 Tests
Catalog # ANTIA74529-96
Supplier: ANTIBODIES.COM