ContactUsLinkComponent
Human Inhibin alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Inhibin alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 48 Tests
Catalog # ANTIA8145-48
Supplier:  ANTIBODIES.COM
New Product
Human Inhibin alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Inhibin alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 48 Tests
Catalog # ANTIA8145-48
Supplier:  ANTIBODIES.COM