Mouse Cytokeratin 18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Cytokeratin 18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 96 Tests
Catalog # ANTIA7029-96
Supplier: Antibodies.com