Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D(-)-Isoleucine

D(-)-Isoleucine

Bottle
Catalog # APOSOR36341-25G
Supplier:   Apollo Scientific
D(-)-Isoleucine
Catalog # APOSOR36341-25G
Supplier:  Apollo Scientific
CAS Number:  319-78-8
D(-)-Isoleucine

Bottle

CAS Number:  319-78-8
Price
SFr. 1’600.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (2R,3R)-(-)-Isoleucine, H-D-Ile-OH
  • CAS Number:319-78-8
  • Formula:H₃CCH₂CH(CH₃)CH(NH₂)COOH
  • MDL Number:MFCD00064221
  • Molecular Weight:131.17 g/mol
  • Melting Pt:265…274 °C
  •  :Density: 1.224 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.