ContactUsLinkComponent
Pure Water Density Standard ISO/IEC 17025 and ISO 17034 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15° 1 * 60 mL
Catalog # ACSDPWD.L06
Supplier:  CPACHEM
New Product
Pure Water Density Standard ISO/IEC 17025 and ISO 17034 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15° 1 * 60 mL
Catalog # ACSDPWD.L06
Supplier:  CPACHEM