Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Penicillamine

D-(-)-Penicillamine

Bottle
Catalog # APOSOR18365-500G
Irritant
Supplier:   Apollo Scientific
D-(-)-Penicillamine
Catalog # APOSOR18365-500G
Supplier:  Apollo Scientific
CAS Number:  52-67-5
D-(-)-Penicillamine

Bottle

CAS Number:  52-67-5
Price
SFr. 790.00
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
  • CAS Number:52-67-5
  • Formula:C₅H₁₁NO₂S
  • MDL Number:MFCD00064302
  • Molecular Weight:149.2 g/mol
  • Melting Pt:∼ 212 °C (dec.)
  • Melting Pt:ca 212 °C (dec.)
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.