Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pyrocatechol 99%

Pyrocatechol 99%

Bottle
Catalog # APOSOR923744-5G
Supplier:   Apollo Scientific
Pyrocatechol 99%
Catalog # APOSOR923744-5G
Supplier:  Apollo Scientific
CAS Number:  120-80-9
Pyrocatechol 99%

Bottle

CAS Number:  120-80-9
Price
SFr. 31.20
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Boiling Pt:245 °C
  • Melting Pt:103…106 °C
  • Flash Pt:131 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.