Order Entry
Switzerland
Orders LinkContactUsLinkComponent
L(+)-Cysteine

L(+)-Cysteine

Bottle
Catalog # APOSOR0325-10KG
Irritant
New Product
Supplier:   Apollo Scientific
L(+)-Cysteine
Catalog # APOSOR0325-10KG
Supplier:  Apollo Scientific
New Product
CAS Number:  52-90-4
L(+)-Cysteine

Bottle

CAS Number:  52-90-4
Price
SFr. 1’100.00
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS Number:52-90-4
  • Formula:HSCH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Melting Pt:240 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-158-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications