Copper 99,9%, nanopowder 30 - 50 nm
Bottle
Catalog # APOSIN1526-100G
Supplier: Apollo Scientific
Copper 99,9%, nanopowder 30 - 50 nm
Catalog # APOSIN1526-100G
Supplier: Apollo Scientific
CAS Number: 7440-50-8
Jump to:
Product Details & Documents
Usage: Inorganic salt or catalyst
- CAS Number:7440-50-8
- Formula:Cu
- MDL Number:MFCD00010965
- Molecular Weight:63,546 g/mol
- UN:3089
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



