Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

D-(-)-Serine

Bottle
Catalog # APOSBIS2001-10KG
Irritant
Supplier:   Apollo Scientific
D-(-)-Serine
Catalog # APOSBIS2001-10KG
Supplier:  Apollo Scientific
CAS Number:  312-84-5
D-(-)-Serine

Bottle

CAS Number:  312-84-5
Price
600.00 SFr.
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  (R)-(-)-Serine, H-D-Ser-OH
  • CAS Number:312-84-5
  • Formula:C₃H₇NO₃
  • MDL Number:MFCD00004269
  • Molecular Weight:105.09 g/mol
  • Melting Pt:220 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-229-4
Specifications

Specifications

  • Pack type:
    Bottle

Specifications