Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Chlorophenol red

Bottle
Catalog # APOSBI4558-25G
Irritant
Supplier:   Apollo Scientific
Chlorophenol red
Catalog # APOSBI4558-25G
Supplier:  Apollo Scientific
CAS Number:  4430-20-0
Chlorophenol red

Bottle

CAS Number:  4430-20-0
Price
SFr. 381.00
Log in for availability
1 * 25 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
  • CAS Number:4430-20-0
  • Formula:C₁₉H₁₂Cl₂O₅S
  • MDL Number:MFCD00005877
  • Molecular Weight:423,273 g/mol
  • EINECS:224-619-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.