ContactUsLinkComponent
1 component: Chromium (VI) (Cr 6+) 18.7mg/l in Water CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 50 mL
Catalog # CMPLRM096851L05
New Product
1 component: Chromium (VI) (Cr 6+) 18.7mg/l in Water CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 50 mL
Catalog # CMPLRM096851L05