ContactUsLinkComponent
Compression spring 0.80 x De 12.00 x L0 22.60nt 5.50, right, 1.4310 stainlessEnds ground and ground Spring unset 1 * 1 ST
Katalognr. MSPP54/6/1
Compression spring 0.80 x De 12.00 x L0 22.60nt 5.50, right, 1.4310 stainlessEnds ground and ground Spring unset 1 * 1 ST
Katalognr. MSPP54/6/1
Frequently Bought Together