ContactUsLinkComponent
12 components: Aluminium (Al) 100mg/l Calcium (Ca) 300mg/l Iron (Fe) 250mg/l Magnesium (Mg) 100mg/l Sodium (Na) 100mg/l Phosphorus (P) 250mg/l Potassium (K) 100mg/l Sulphur (S) 100mg/l Carbon (C) 200mg/l Chlorides (Cl-) 2000mg/l M 1 * 100 mL
Catalog # CMPLRM023314L1
New Product
12 components: Aluminium (Al) 100mg/l Calcium (Ca) 300mg/l Iron (Fe) 250mg/l Magnesium (Mg) 100mg/l Sodium (Na) 100mg/l Phosphorus (P) 250mg/l Potassium (K) 100mg/l Sulphur (S) 100mg/l Carbon (C) 200mg/l Chlorides (Cl-) 2000mg/l M 1 * 100 mL
Catalog # CMPLRM023314L1