ContactUsLinkComponent
Sodium Carbonate 0.02M (0.02N) CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1.000 mL
Catalog # CMPLZ15034583.1L
New Product
Sodium Carbonate 0.02M (0.02N) CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1.000 mL
Catalog # CMPLZ15034583.1L