ContactUsLinkComponent
Mini-Display Leak-Sensor, Alarm box in plastic housing for an installation in front plates of lab furniture, front with touch-buttons and LED indicators, exclusively programmed to control a b.safe Splitter and to monitor 2 containers for automatic sw 1 * 1 items
Catalog # BOHLR658-24
Supplier:  Bohlender
New Product
Mini-Display Leak-Sensor, Alarm box in plastic housing for an installation in front plates of lab furniture, front with touch-buttons and LED indicators, exclusively programmed to control a b.safe Splitter and to monitor 2 containers for automatic sw 1 * 1 items
Catalog # BOHLR658-24
Supplier:  Bohlender