1 component: F3CCOOH 100ug/ml in CH4O / H2O (1/1) CRM, ISO 17034 and ISO 17025 Conservation 24 months. Store in a refrigerator at temperatures between 2°C to 8°C 1 * 1 mL
Catalog # CMPLRM1179201
Supplier: CUSTOM MADE CHEMICALS LAB