[EN]LEUKOSILK 5 M X 2.50 CM 1 * 1 ST
Catalog # SOEH1009210
Supplier: SOEHNGEN
New Product
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Supplier: EPPENDORF
Nitrile gloves, Kimtech™ Blue Nitrile™
Supplier: KIMBERLY CLARK
Detergents for glassware washers, ProCare Lab 10 AP
Supplier: MIELE