ContactUsLinkComponent
Hamilton Star and Vantage compatible P50 Tips, non-sterile working volume 50 uL non-sterile (RNAse/DNAse free) clear tips compare to Hamilton 235446 Note: NTR style 5 x 4 stack with 384 tips per stack [7.680 tips] 1 * 7.680 items
Catalog # FLUTHAM-50.NS
Supplier:  FLUOTICS
New Product
Hamilton Star and Vantage compatible P50 Tips, non-sterile working volume 50 uL non-sterile (RNAse/DNAse free) clear tips compare to Hamilton 235446 Note: NTR style 5 x 4 stack with 384 tips per stack [7.680 tips] 1 * 7.680 items
Catalog # FLUTHAM-50.NS
Supplier:  FLUOTICS