Insert E 139/1 For the optimum loading of test tubes up to 12 x 200 mm 1 * 1 items
Catalog # MIEL12667390
Supplier: MIELE
New Product
Insert E 139/1 For the optimum loading of test tubes up to 12 x 200 mm 1 * 1 items
Catalog # MIEL12667390
Supplier: MIELE
New Product
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Silicone Lab Mat
Supplier: VWR Collection
Benzene, Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals