ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 2x7=14 plates, 90mm high , stainless steel, dimensions (HxDxW) 185x630x13 5mm 1 * 1 items
Catalog # TENATE35678
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 2x7=14 plates, 90mm high , stainless steel, dimensions (HxDxW) 185x630x13 5mm 1 * 1 items
Catalog # TENATE35678
Supplier:  TENAK