ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 2x6=12 plates, 90mm high , stainless steel, dimensions (HxDxW) 185x540x13 5mm 1 * 1 items
Catalog # TENATE35672
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 2x6=12 plates, 90mm high , stainless steel, dimensions (HxDxW) 185x540x13 5mm 1 * 1 items
Catalog # TENATE35672
Supplier:  TENAK