ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 3x6=18 plates, 55mm high , stainless steel, dimensions (HxDxW) 172x540x13 5mm 1 * 1 items
Catalog # TENATE35638
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 3x6=18 plates, 55mm high , stainless steel, dimensions (HxDxW) 172x540x13 5mm 1 * 1 items
Catalog # TENATE35638
Supplier:  TENAK