ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 11x7=77 plates, 35mm high , stainless steel, dimensions (HxDxW) 408x630x1 35mm 1 * 1 items
Catalog # TENATE35346
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 11x7=77 plates, 35mm high , stainless steel, dimensions (HxDxW) 408x630x1 35mm 1 * 1 items
Catalog # TENATE35346
Supplier:  TENAK