ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 7x7=49 plates, 35mm high , stainless steel, dimensions (HxDxW) 260x630x13 5mm 1 * 1 items
Catalog # TENATE35338
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 7x7=49 plates, 35mm high , stainless steel, dimensions (HxDxW) 260x630x13 5mm 1 * 1 items
Catalog # TENATE35338
Supplier:  TENAK