ContactUsLinkComponent
Comfort MTP rack for upright freezers for SBS format (HxD) 6x5=30 plates, 35mm high , stainless steel, dimensions (HxDxW) 223x450x13 5mm 1 * 1 items
Catalog # TENATE35312
Supplier:  TENAK
New Product
Comfort MTP rack for upright freezers for SBS format (HxD) 6x5=30 plates, 35mm high , stainless steel, dimensions (HxDxW) 223x450x13 5mm 1 * 1 items
Catalog # TENATE35312
Supplier:  TENAK