ContactUsLinkComponent
Comfort rack for chest freezers for 13 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, with spring lock, dimens ions (HxDxW) 466x135x92mm 1 * 1 items
Catalog # TENATE30530
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 13 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, with spring lock, dimens ions (HxDxW) 466x135x92mm 1 * 1 items
Catalog # TENATE30530
Supplier:  TENAK