ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 7x4=28 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, sliding shel ves, dimensions (HxDxW) 253x367x135mm 1 * 1 items
Catalog # TENATE30224
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 7x4=28 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, sliding shel ves, dimensions (HxDxW) 253x367x135mm 1 * 1 items
Catalog # TENATE30224
Supplier:  TENAK