ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 5x5=25 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, dimensions (HxDxW) 181x452x135mm 1 * 1 items
Catalog # TENATE30130
Supplier:  TENAK
New Product
Standard rack for upright freezers for (HxD) 5x5=25 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, dimensions (HxDxW) 181x452x135mm 1 * 1 items
Catalog # TENATE30130
Supplier:  TENAK