ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 4x7=28 plates, 90mm high, stainless steel, dimensions (HxDxW) 369x630x13 5mm 1 * 1 items
Catalog # TENATE27682
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 4x7=28 plates, 90mm high, stainless steel, dimensions (HxDxW) 369x630x13 5mm 1 * 1 items
Catalog # TENATE27682
Supplier:  TENAK