ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 4x6=24 plates, 55mm high, stainless steel, dimensions (HxDxW) 229x540x13 5mm 1 * 1 items
Catalog # TENATE27640
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 4x6=24 plates, 55mm high, stainless steel, dimensions (HxDxW) 229x540x13 5mm 1 * 1 items
Catalog # TENATE27640
Supplier:  TENAK