ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 4x4=16 plates, 55mm high, stainless steel, dimensions (HxDxW) 229x360x13 5mm 1 * 1 items
Catalog # TENATE27622
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 4x4=16 plates, 55mm high, stainless steel, dimensions (HxDxW) 229x360x13 5mm 1 * 1 items
Catalog # TENATE27622
Supplier:  TENAK