ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 3x4=12 plates, 55mm high, stainless steel, dimensions (HxDxW) 172x360x13 5mm 1 * 1 items
Catalog # TENATE27620
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 3x4=12 plates, 55mm high, stainless steel, dimensions (HxDxW) 172x360x13 5mm 1 * 1 items
Catalog # TENATE27620
Supplier:  TENAK