ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 15x6=90 plates, 25mm high, stainless steel, dimensions (HxDxW) 406x540x1 35mm 1 * 1 items
Catalog # TENATE27539
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 15x6=90 plates, 25mm high, stainless steel, dimensions (HxDxW) 406x540x1 35mm 1 * 1 items
Catalog # TENATE27539
Supplier:  TENAK