ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 7x6=42 plates, 35mm high, stainless steel, dimensions (HxDxW) 260x540x13 5mm 1 * 1 items
Catalog # TENATE27326
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 7x6=42 plates, 35mm high, stainless steel, dimensions (HxDxW) 260x540x13 5mm 1 * 1 items
Catalog # TENATE27326
Supplier:  TENAK