ContactUsLinkComponent
Standard MTP rack for upright freezers for SBS format (HxD) 6x6=36 plates, 35mm high, stainless steel, dimensions (HxDxW) 223x540x13 5mm 1 * 1 items
Catalog # TENATE27324
Supplier:  TENAK
New Product
Standard MTP rack for upright freezers for SBS format (HxD) 6x6=36 plates, 35mm high, stainless steel, dimensions (HxDxW) 223x540x13 5mm 1 * 1 items
Catalog # TENATE27324
Supplier:  TENAK