ContactUsLinkComponent
Standard MTP racks for chest freezers for 13 SBS format plates, 35mm high, stainless steel, dimensions (HxDxW) 479x135x92mm 1 * 1 items
Catalog # TENATE27240
Supplier:  TENAK
New Product
Standard MTP racks for chest freezers for 13 SBS format plates, 35mm high, stainless steel, dimensions (HxDxW) 479x135x92mm 1 * 1 items
Catalog # TENATE27240
Supplier:  TENAK