ContactUsLinkComponent
Comfort rack for chest freezers for 2 cryoboxes, 130mm high, stainless steel, with spring lock, dimensions (HxDxW) 635x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE25470B
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 2 cryoboxes, 130mm high, stainless steel, with spring lock, dimensions (HxDxW) 635x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE25470B
Supplier:  TENAK