ContactUsLinkComponent
Comfort rack for upright freezers for (HxD) 2x5=10 cryoboxes, 130mm high, stainless steel, sliding shelves, dimensions (HxDxW) 268x6 83x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE25206B
Supplier:  TENAK
New Product
Comfort rack for upright freezers for (HxD) 2x5=10 cryoboxes, 130mm high, stainless steel, sliding shelves, dimensions (HxDxW) 268x6 83x140mm, delivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE25206B
Supplier:  TENAK