ContactUsLinkComponent
Classic rack for chest freezers for 5 cryoboxes, 130mm high, stainless steel, with locking rod, dimensions (HxDxW) 669x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE25106B
Supplier:  TENAK
New Product
Classic rack for chest freezers for 5 cryoboxes, 130mm high, stainless steel, with locking rod, dimensions (HxDxW) 669x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE25106B
Supplier:  TENAK