ContactUsLinkComponent
Comfort rack for chest freezers for 4 cryoboxes, 100mm high, stainless steel, with spring lock, dimensions (HxDxW) 420x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24562B10
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 4 cryoboxes, 100mm high, stainless steel, with spring lock, dimensions (HxDxW) 420x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE24562B10
Supplier:  TENAK