ContactUsLinkComponent
Comfort rack for chest freezers for 3 cryoboxes, 96mm high, stainless steel, with spring lock, dimensions (HxDxW) 305x140x140mm 1 * 1 items
Catalog # TENATE24560L
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 3 cryoboxes, 96mm high, stainless steel, with spring lock, dimensions (HxDxW) 305x140x140mm 1 * 1 items
Catalog # TENATE24560L
Supplier:  TENAK