ContactUsLinkComponent
Comfort rack for chest freezers for 5 cryoboxes, 50mm high, stainless steel, with spring lock, dimensions (HxDxW) 280x83x83mm 1 * 1 items
Catalog # TENATE24486
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 5 cryoboxes, 50mm high, stainless steel, with spring lock, dimensions (HxDxW) 280x83x83mm 1 * 1 items
Catalog # TENATE24486
Supplier:  TENAK